Assay Details
Target Gene Details
Entrez Gene ID: | 57260 |
Gene Name: | leukotriene B4 receptor 2 |
Gene Aliases: |
5830462O07Rik, BLT2, LTB4-R 2, LTB4-R-2, LTB4-R2 |
Location: |
Chr.14:55757916-55764066 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltb4r2 | NM_020490.2 | 2 | 859 | NP_065236.1 |
| AB029893.1 | 1 | 726 | BAA99554.1 | |
| AK018029.1 | 2 | 860 | ||
| BC106834.2 | 1 | 726 | AAI06835.1 | |
| BC106835.2 | 1 | 726 | AAI06836.1 |
More Information
Set Membership: |
|

Genomic Map