Assay Details
Target Gene Details
Entrez Gene ID: | 70069 |
Gene Name: | H1 histone family, member N, testis-specific |
Gene Aliases: |
1700026P10Rik, H1.7, H1t2, Hanp1, mH1t2 |
Location: |
Chr.15:98255982-98257307 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H1fnt | NM_027304.2 | 1 | 900 | NP_081580.2 |
| AB016273.1 | 1 | 872 | BAC20949.1 | |
| AK006402.1 | 1 | 639 | BAB24570.1 | |
| AY496853.1 | 1 | 898 | AAS49491.1 | |
| BC052363.1 | 1 | 886 | AAH52363.1 |
More Information
Set Membership: |
|

Genomic Map