Assay Details
Target Gene Details
Entrez Gene ID: | 68942 |
Gene Name: | charged multivesicular body protein 2B |
Gene Aliases: |
1190006E07Rik |
Location: |
Chr.16:65539133-65562697 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp2b | NM_026879.2 | 6 | 1092 | NP_081155.1 |
| AK004506.1 | 3 | 565 | BAB23339.1 | |
| AK084111.1 | 6 | 1094 | BAC39118.1 | |
| AK181292.1 | 1 | 216 | ||
| AK201890.1 | 1 | 216 | ||
| AK206751.1 | 1 | 216 | ||
| AK208493.1 | 1 | 216 | ||
| AK209621.1 | 1 | 216 | ||
| AK214190.1 | 1 | 216 | ||
| AK215317.1 | 1 | 216 | ||
| AK217623.1 | 1 | 216 | ||
| AK218468.1 | 1 | 216 | ||
| BC042626.1 | 5 | 848 | AAH42626.1 | |
| BC055809.1 | 6 | 1121 | AAH55809.1 |
More Information
Set Membership: |
|

Genomic Map