Assay Details
Target Gene Details
Entrez Gene ID: | 105787 |
Gene Name: | protein kinase, AMP-activated, alpha 1 catalytic subunit |
Gene Aliases: |
AI194361, AI450832, AL024255, AMPKalpha1, C130083N04Rik |
Location: |
Chr.15:5143861-5181899 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkaa1 | NM_001013367.3 | 7 | 1073 | NP_001013385.3 |
| AK147811.1 | 7 | 1050 | ||
| AK160612.1 | 7 | 1054 | BAE35914.1 | |
| AY885266.1 | 7 | 1032 | AAW79567.1 | |
| BC086695.1 | 7 | 1043 | ||
| CB245934.1 | 2 | 369 |
More Information
Set Membership: |
|

Genomic Map