Assay Details
Target Gene Details
Entrez Gene ID: | 51789 |
Gene Name: | tyrosine kinase, non-receptor, 2 |
Gene Aliases: |
Ack, Ack-1, Ack1, Cdgip, Pyk1 |
Location: |
Chr.16:32644099-32683493 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnk2 | NM_001110147.1 | NP_001103617.1 | ||
| NM_016788.3 | NP_058068.2 | |||
| AF037260.1 | AAC04786.1 | |||
| BC052421.1 | AAH52421.1 | |||
| DQ666696.1 |
Target Gene Details
Entrez Gene ID: | 675921 |
Gene Name: | tyrosine kinase, non-receptor 2, opposite strand |
Gene Aliases: |
ENSMUSG00000075265, Gm10818 |
Location: |
Chr.16:32665223-32668197 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnk2os | NR_033493.1 | 2 | 457 | |
| AK153635.1 | 2 | 458 | BAE32130.1 |
More Information
Set Membership: |
|

Genomic Map