Assay Details
Target Gene Details
Entrez Gene ID: | 14702 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 2 |
Gene Aliases: |
82 |
Location: |
Chr.14:19872558-19977462 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng2 | NM_001038637.1 | 5 | 2552 | NP_001033726.1 |
| NM_001285908.1 | 4 | 2512 | NP_001272837.1 | |
| NM_001285909.1 | 6 | 2619 | NP_001272838.1 | |
| NM_001285910.1 | 3 | 2659 | NP_001272839.1 | |
| NM_001285911.1 | 4 | 2527 | NP_001272840.1 | |
| NM_010315.4 | 4 | 2700 | NP_034445.1 | |
| AK003394.1 | 1 | 220 | ||
| AK075698.1 | 4 | 2430 | BAC35896.1 | |
| AK148899.1 | 1 | 1080 | ||
| AK149133.1 | 1 | 628 | ||
| AK158697.1 | 4 | 2701 | BAE34615.1 | |
| AK160396.1 | 4 | 2430 | BAE35765.1 |
More Information
Set Membership: |
|

Genomic Map