Assay Details
Target Gene Details
Entrez Gene ID: | 105651 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3E |
Gene Aliases: |
A630071A11Rik, AW049239, Gm1211 |
Location: |
Chr.14:54875597-54877538 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3e | NM_001167908.1 | 1 | 185 | NP_001161380.1 |
| BB645735.1 | 1 | 186 |
More Information
Set Membership: |
|

Genomic Map