Assay Details
Target Gene Details
Entrez Gene ID: | 15424 |
Gene Name: | homeobox C5 |
Gene Aliases: |
AI181837, Hox-3.4, Hox-6.2 |
Location: |
Chr.15:103014008-103017429 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxc5 | NM_175730.5 | 2 | 897 | NP_783857.1 |
| AK078527.1 | 2 | 898 | BAC37324.1 | |
| BC115551.1 | 2 | 898 | ||
| BC115552.1 | 2 | 898 |
More Information
Set Membership: |
|

Genomic Map