Assay Details
Target Gene Details
Entrez Gene ID: | 15425 |
Gene Name: | homeobox C6 |
Gene Aliases: |
Hox-3.3, Hox-6.1 |
Location: |
Chr.15:102998226-103011882 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxc6 | NM_010465.2 | 1 | 364 | NP_034595.2 |
| AB221685.1 | 1 | 320 | ||
| AK050798.1 | 1 | 132 | BAC34415.1 | |
| BC141061.1 | 1 | 343 | ||
| BX528590.1 | 1 | 365 | ||
| X12504.1 | 2 | 389 | CAA31022.1 | |
| X16511.1 | 1 | 447 | CAA34518.1 |
More Information
Set Membership: |
|

Genomic Map