Assay Details
Target Gene Details
Entrez Gene ID: | 15425 |
Gene Name: | homeobox C6 |
Gene Aliases: |
Hox-3.3, Hox-6.1 |
Location: |
Chr.15:102998226-103011882 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxc6 | NM_010465.2 | 2 | 826 | NP_034595.2 |
| AK050798.1 | 2 | 594 | BAC34415.1 | |
| BC141061.1 | 2 | 805 | ||
| J03074.1 | 1 | 386 | AAA37832.1 | |
| X12504.1 | 3 | 851 | CAA31022.1 | |
| X16511.1 | 2 | 909 | CAA34518.1 |
More Information
Set Membership: |
|

Genomic Map