Assay Details
Target Gene Details
Entrez Gene ID: | 20788 |
Gene Name: | sterol regulatory element binding factor 2 |
Gene Aliases: |
AI608257, SREBP-2, SREBP2, SREBP2gc, bHLHd2, lop13, nuc |
Location: |
Chr.15:82147269-82205379 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srebf2 | NM_033218.1 | 10 | 1953 | NP_150087.1 |
| AF374267.1 | 10 | 1806 | AAK54763.1 | |
| AK155857.1 | 11 | 1901 | BAE33463.1 | |
| AK172219.1 | 10 | 1960 | ||
| AK192672.1 | ||||
| BQ963055.1 |

Genomic Map