Assay Details
Target Gene Details
Entrez Gene ID: | 64833 |
Gene Name: | acyl-CoA thioesterase 10 |
Gene Aliases: |
Acate3, MT-ACT48, p48 |
Location: |
Chr.15:20665214-20666750 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot10 | NM_022816.2 | 1 | 267 | NP_073727.2 |
| AJ238894.1 | 1 | 267 | CAB45255.1 | |
| BC108959.1 | 1 | 258 | AAI08960.1 | |
| BC108960.1 | 1 | 258 | AAI08961.1 |
Target Gene Details
Entrez Gene ID: | 215654 |
Gene Name: | cadherin 12 |
Gene Aliases: |
Cdhb |
Location: |
Chr.15:20448879-21597005 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh12 |
More Information
Set Membership: |
|

Genomic Map