Assay Details
Target Gene Details
Entrez Gene ID: | 16421 |
Gene Name: | integrin beta 7 |
Gene Aliases: |
Ly69 |
Location: |
Chr.15:102215994-102231958 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb7 | NM_013566.2 | 15 | 2580 | NP_038594.2 |
| AI322899.1 | 2 | 253 | ||
| AK078871.1 | 15 | 2581 | BAC37433.1 | |
| AK144773.1 | 15 | 2555 | ||
| AK154766.1 | 15 | 2581 | BAE32815.1 | |
| AK155207.1 | 15 | 2547 | BAE33119.1 | |
| AK155594.1 | 15 | 2553 | ||
| BC011184.1 | 15 | 2553 | AAH11184.1 | |
| M68903.1 | 15 | 2528 | AAA02749.1 | |
| M95632.1 | 15 | 2524 | AAA39323.1 | |
| M95633.1 | 15 | 2524 | AAA39324.1 |
More Information
Set Membership: |
|

Genomic Map