Assay Details
Target Gene Details
Entrez Gene ID: | 105837 |
Gene Name: | Mdm2, transformed 3T3 cell double minute p53 binding protein |
Gene Aliases: |
AI429604, MDM2BP |
Location: |
Chr.15:55557408-55626423 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtbp | NM_134092.3 | 22 | 2957 | NP_598853.3 |
| AJ278508.1 | 22 | 2878 | CAC07970.1 | |
| AK080036.1 | 22 | 2771 | BAC37810.1 | |
| AK088652.1 | 12 | 2385 | BAC40479.1 | |
| AK133855.1 | 22 | 2805 | BAE21889.1 | |
| BC005500.1 | 15 | 1954 | AAH05500.1 | |
| BC052197.1 | 22 | 2957 | AAH52197.1 |

Genomic Map