Assay Details
Target Gene Details
Entrez Gene ID: | 66194 |
Gene Name: | pyrroline-5-carboxylate reductase-like |
Gene Aliases: |
1110058B13Rik, 2700073G24Rik |
Location: |
Chr.15:75916463-75921560 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pycrl | NM_025412.2 | 6 | 939 | NP_079688.2 |
| AK002912.1 | 6 | 870 | BAB22451.1 | |
| AK004291.1 | 6 | 943 | BAB23252.1 | |
| AK013214.1 | 6 | 864 | ||
| BC026536.1 | 6 | 860 | AAH26536.1 | |
| BF721406.1 | 1 | 415 |
More Information
Set Membership: |
|

Genomic Map