Assay Details
Target Gene Details
Entrez Gene ID: | 20024 |
Gene Name: | SUB1 homolog (S. cerevisiae) |
Gene Aliases: |
AI842364, P15, P9, Pc4, Rpo2tc1 |
Location: |
Chr.15:11981338-11996031 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sub1 | NM_011294.3 | 5 | 857 | NP_035424.1 |
| AK048371.1 | 5 | 857 | BAC33314.1 | |
| AK049260.1 | 5 | 857 | BAC33642.1 | |
| J03750.1 | 5 | 841 | AAA37317.1 |
More Information
Set Membership: |
|

Genomic Map