Assay Details
Target Gene Details
Entrez Gene ID: | 22762 |
Gene Name: | zinc finger protein, multitype 2 |
Gene Aliases: |
B330005D23Rik, FOG-2, FOG2 |
Location: |
Chr.15:40655042-41104592 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfpm2 | NM_011766.5 | 8 | 2636 | NP_035896.1 |
| AF107306.1 | 8 | 2157 | AAD12182.1 | |
| AF118845.1 | 8 | 2406 | AAD17251.1 | |
| AF125166.1 | 8 | 2637 | AAD33250.1 | |
| BC059241.1 | 9 | 2482 | AAH59241.1 | |
| EF514219.1 | 8 | 3459 |
More Information
Set Membership: |
|

Genomic Map