Assay Details
Target Gene Details
Entrez Gene ID: | 71756 |
Gene Name: | carboxypeptidase N, polypeptide 2 |
Gene Aliases: |
1300018K11Rik |
Location: |
Chr.16:30256378-30267614 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpn2 | NM_027904.3 | 2 | 2330 | NP_082180.2 |
| AK005049.1 | BAB23775.1 | |||
| BC025836.1 | 2 | 2297 | AAH25836.1 | |
| BC081550.1 | 1 | 1849 | AAH81550.1 |
More Information
Set Membership: |
|

Genomic Map