Assay Details
Target Gene Details
Entrez Gene ID: | 16419 |
Gene Name: | integrin beta 5 |
Gene Aliases: |
AA475909, AI874634, ESTM23, [b]-5, [b]5, [b]5A, [b]5B, beta-5, beta5 |
Location: |
Chr.16:33829665-33949338 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb5 | NM_001145884.1 | 1 | 124 | NP_001139356.1 |
| NM_010580.2 | 1 | 124 | NP_034710.2 | |
| AF022110.1 | 1 | 124 | AAD08782.1 | |
| AF043256.1 | AAC40109.1 | |||
| AF043257.1 | AAC40110.1 | |||
| AK140251.1 | BAE24299.1 | |||
| AK143532.1 | 1 | 58 | ||
| BC046280.1 | ||||
| BC058246.1 | 1 | 88 | AAH58246.1 |
More Information
Set Membership: |
|

Genomic Map