Assay Details
Target Gene Details
Entrez Gene ID: | 21974 |
Gene Name: | topoisomerase (DNA) II beta |
Gene Aliases: |
D230016L12Rik, Top-2 |
Location: |
Chr.14:16365206-16430787 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top2b | NM_009409.2 | 1 | 79 | NP_033435.2 |
| AK041054.1 | 1 | 106 | BAC30801.1 | |
| AK087535.1 | 1 | 108 | ||
| AK163043.1 | 1 | 108 | BAE37168.1 | |
| BC041106.1 | 1 | 99 | AAH41106.1 | |
| BC054541.1 | 1 | 79 | AAH54541.1 |
More Information
Set Membership: |
|

Genomic Map