Assay Details
Target Gene Details
Entrez Gene ID: | 380959 |
Gene Name: | asparagine-linked glycosylation 10B (alpha-1,2-glucosyltransferase) |
Gene Aliases: |
AA469671, AU045404, AW123895, C85569, Deaf1, Gm917, nse5 |
Location: |
Chr.15:90224311-90230554 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alg10b | NM_001033441.3 | 3 | 3181 | NP_001028613.1 |
| AK147821.1 | 3 | 3182 | BAE28159.1 | |
| AK153580.1 | 3 | 3012 | ||
| AK162193.1 | 1 | 536 | ||
| AK170450.1 | 3 | 3182 | ||
| AK171179.1 | 3 | 2994 |
More Information
Set Membership: |
|

Genomic Map