Assay Details
Target Gene Details
Entrez Gene ID: | 105827 |
Gene Name: | adhesion molecule with Ig like domain 2 |
Gene Aliases: |
AI415330, AMIGO-2, AW208913, Ali1 |
Location: |
Chr.15:97244074-97247287 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amigo2 | NM_001164563.1 | 2 | 821 | NP_001158035.1 |
| NM_001164602.1 | 2 | 789 | NP_001158074.1 | |
| NM_178114.4 | 1 | 1164 | NP_835215.1 | |
| AB078880.1 | 1 | 822 | BAC81187.1 | |
| AK030848.1 | 1 | 1131 | ||
| AK038783.1 | 2 | 822 | ||
| AY237006.1 | 1 | 416 | AAO48947.1 | |
| BC095990.1 | 1 | 999 | AAH95990.1 |

Genomic Map