Assay Details
Target Gene Details
Entrez Gene ID: | 208177 |
Gene Name: | pleckstrin homology-like domain, family B, member 2 |
Gene Aliases: |
AV253284, C820004H04Rik, LL5b, LL5beta |
Location: |
Chr.16:45746230-45953701 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phldb2 | AK050493.1 |

Genomic Map