Assay Details
Target Gene Details
Entrez Gene ID: | 223693 |
Gene Name: | transmembrane protein 184b |
Gene Aliases: |
2610507A11, 4732495E13Rik |
Location: |
Chr.15:79360684-79403303 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem184b | NM_001253817.1 | 9 | 2943 | NP_001240746.1 |
| NM_001253819.1 | 10 | 2985 | NP_001240748.1 | |
| NM_001253820.1 | 7 | 2509 | NP_001240749.1 | |
| NM_172608.1 | 9 | 2989 | NP_766196.1 | |
| AA240987.1 | ||||
| AK028196.1 | 9 | 2947 | BAC25805.1 | |
| AK029129.1 | 9 | 2967 | BAC26312.1 | |
| AK048138.1 | 10 | 2964 | BAC33254.1 | |
| AK149366.1 | 9 | 2970 | BAE28837.1 | |
| AK158789.1 | 9 | 2990 | BAE34666.1 | |
| BC026687.1 | 1 | 1481 | AAH26687.1 | |
| BC046959.1 | 9 | 2943 | AAH46959.1 |
More Information
Set Membership: |
|

Genomic Map