Assay Details
Target Gene Details
Entrez Gene ID: | 71753 |
Gene Name: | transmembrane serine protease 6 |
Gene Aliases: |
1300008A22Rik |
Location: |
Chr.15:78439667-78468648 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss6 | NM_027902.2 | 18 | 2718 | NP_082178.2 |
| AK004939.2 | 18 | 2568 | BAB23684.2 | |
| AY240929.1 | 18 | 2695 | AAP69827.1 | |
| BC029645.1 | 18 | 2695 | AAH29645.2 | |
| BC057674.1 | 18 | 2660 | AAH57674.1 |
More Information
Set Membership: |
|

Genomic Map