Assay Details
Target Gene Details
Entrez Gene ID: | 109754 |
Gene Name: | cytochrome b5 reductase 3 |
Gene Aliases: |
0610016L08Rik, 2500002N19Rik, C85115, Dia-1, Dia1, WU:AL591952.1-001, WU:AL591952.1-002, WU:AL591952.1-003, WU:Cyb5r3 |
Location: |
Chr.15:83153495-83172208 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyb5r3 | NM_029787.2 | 9 | 1632 | NP_084063.1 |
| AK002640.1 | 9 | 1604 | BAB22252.1 | |
| AK128917.1 | 9 | 1626 | ||
| AK218164.1 | 1 | 275 | ||
| AK218827.1 | 1 | 292 | ||
| BC004760.1 | 9 | 1639 | AAH04760.1 | |
| BC032013.1 | 9 | 1614 | AAH32013.1 | |
| BC043074.1 | 9 | 1632 | AAH43074.1 |
More Information
Set Membership: |
|

Genomic Map