Assay Details
Target Gene Details
Entrez Gene ID: | 11877 |
Gene Name: | armadillo repeat gene deleted in velo-cardio-facial syndrome |
Gene Aliases: |
- |
Location: |
Chr.16:18348182-18407076 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arvcf | NM_001272028.1 | 18 | 4325 | NP_001258957.1 |
| NM_001272029.1 | 18 | 4458 | NP_001258958.1 | |
| NM_001272030.1 | 17 | 4307 | NP_001258959.1 | |
| NM_001272031.1 | 17 | 4068 | NP_001258960.1 | |
| NM_001272032.1 | 16 | 4050 | NP_001258961.1 | |
| NM_033474.3 | 19 | 4476 | NP_258435.2 | |
| AK044982.1 | 16 | 3998 | BAC32169.1 | |
| BC051655.1 | 13 | 2473 | AAH51655.1 | |
| BC056980.1 | 19 | 4384 | AAH56980.1 |
More Information
Set Membership: |
|

Genomic Map