Assay Details
Target Gene Details
Entrez Gene ID: | 382913 |
Gene Name: | nei like 2 (E. coli) |
Gene Aliases: |
Gm1212, NEH2 |
Location: |
Chr.14:63182445-63194244 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neil2 | NM_201610.2 | 5 | 1174 | NP_963904.2 |
| AY518221.1 | 5 | 1190 | AAR98807.1 | |
| BC115709.1 | ||||
| BC115762.1 | AAI15763.1 |
More Information
Set Membership: |
|

Genomic Map