Assay Details
Target Gene Details
Entrez Gene ID: | 67053 |
Gene Name: | ribonuclease P 14 subunit |
Gene Aliases: |
2610511E03Rik, AA682089 |
Location: |
Chr.14:8063265-8092127 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpp14 | NM_025938.4 | 6 | 852 | NP_080214.1 |
| AK012105.1 | 6 | 799 | BAB28036.1 | |
| AK016631.1 | 6 | 799 | BAB30347.1 | |
| BC026988.1 | 6 | 782 | AAH26988.1 | |
| BC046803.1 | 6 | 780 | AAH46803.1 |

Genomic Map