Assay Details
Target Gene Details
Entrez Gene ID: | 18285 |
Gene Name: | outer dense fiber of sperm tails 1 |
Gene Aliases: |
- |
Location: |
Chr.15:38219203-38226735 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Odf1 | NM_008757.3 | 1 | 251 | NP_032783.2 |
| BC145758.1 | 1 | 251 | ||
| CJ042343.1 | 1 | 140 | ||
| EU791469.1 | 1 | 251 | ||
| X79446.1 | 1 | 254 | CAA55965.1 |

Genomic Map