Assay Details
Target Gene Details
Entrez Gene ID: | 22045 |
Gene Name: | thyrotropin releasing hormone receptor |
Gene Aliases: |
TRH-R1 |
Location: |
Chr.15:44195924-44235912 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trhr | NM_013696.2 | 3 | 1401 | NP_038724.1 |
| AK134591.1 | 3 | 1402 | BAE22196.1 | |
| BC109005.1 | 3 | 1286 | AAI09006.1 | |
| BC109006.1 | 3 | 1286 | AAI09007.1 | |
| BC128269.1 | 3 | 1286 | ||
| L48936.1 | 1 | 59 | AAA81559.1 | |
| M59811.1 | 3 | 1310 | AAA40480.1 | |
| M94384.1 | 3 | 1310 | AAA40437.1 |
More Information
Set Membership: |
|

Genomic Map