Assay Details
Target Gene Details
Entrez Gene ID: | 69721 |
Gene Name: | NFKB inhibitor interacting Ras-like protein 1 |
Gene Aliases: |
2400004O09Rik, KBRAS1, kappaB-Ras1 |
Location: |
Chr.14:18271142-18284003 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkiras1 | NM_023526.3 | 4 | 3229 | NP_076015.2 |
| AK148040.1 | 4 | 3155 | BAE28306.1 |

Genomic Map