Assay Details
Target Gene Details
Entrez Gene ID: | 218973 |
Gene Name: | WD repeat and HMG-box DNA binding protein 1 |
Gene Aliases: |
A630092E18, AND-1, D630024B06Rik |
Location: |
Chr.14:47240944-47276857 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdhd1 | NM_172598.3 | 25 | 3448 | NP_766186.2 |
| AK160777.1 | 5 | 609 | BAE36001.1 | |
| BC063740.1 | 25 | 3381 | AAH63740.1 |
More Information
Set Membership: |
|

Genomic Map