Assay Details
Target Gene Details
Entrez Gene ID: | 219149 |
Gene Name: | X Kell blood group precursor related family member 6 homolog |
Gene Aliases: |
BC024502, Xrg6 |
Location: |
Chr.14:63606530-63820411 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xkr6 | NM_173393.2 | NP_775569.2 | ||
| AY534252.1 | AAT07101.1 | |||
| BC024502.1 | 1 | 159 | AAH24502.1 |
More Information
Set Membership: |
|

Genomic Map