Assay Details
Target Gene Details
Entrez Gene ID: | 12224 |
Gene Name: | Kruppel-like factor 5 |
Gene Aliases: |
4930520J07Rik, Bteb2, CKLF, IKLF |
Location: |
Chr.14:99298691-99313412 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf5 | NM_009769.4 | 4 | 1686 | NP_033899.2 |
| AB025099.3 | BAA92284.3 | |||
| AF079852.1 | 4 | 1548 | AAD17696.1 | |
| AK087066.1 | 4 | 1530 | ||
| BC012958.1 | 4 | 1687 | AAH12958.1 |
More Information
Set Membership: |
|

Genomic Map