Assay Details
Target Gene Details
Entrez Gene ID: | 219094 |
Gene Name: | KH and NYN domain containing |
Gene Aliases: |
6330555F21, 9130227C08Rik, AI605259, Kiaa0323, mKIAA0323 |
Location: |
Chr.14:55884964-55898780 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Khnyn | AK076856.1 | 1 | 2233 | |
| AK122247.1 | 8 | 4752 | BAC65529.1 |
Target Gene Details
Entrez Gene ID: | 654795 |
Gene Name: | short chain dehydrogenase/reductase family 39U, member 1 |
Gene Aliases: |
2310014G06Rik |
Location: |
Chr.14:55897285-55900281 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdr39u1 | NM_001082975.1 | 6 | 867 | NP_001076444.1 |
| AK009339.1 | 6 | 847 | BAE43214.1 | |
| AK049709.1 | 6 | 861 | BAC33886.1 | |
| BC087941.1 | AAH87941.1 |
More Information
Set Membership: |
|

Genomic Map