Assay Details
Target Gene Details
Entrez Gene ID: | 628705 |
Gene Name: | PHD finger protein 11C |
Gene Aliases: |
EG628705, Gm6907, Phf11 |
Location: |
Chr.14:59380833-59440890 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf11c |
Target Gene Details
Entrez Gene ID: | 239122 |
Gene Name: | SET domain, bifurcated 2 |
Gene Aliases: |
Clld8, Gm293, KMT1F |
Location: |
Chr.14:59402011-59440877 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setdb2 | NM_001081024.1 | 13 | 2316 | NP_001074493.1 |
| BC157926.1 | 12 | 2138 | ||
| BC172021.1 | 12 | 2138 | ||
| EU155090.1 | 12 | 2130 | ||
| EU155091.1 | 12 | 2130 | ||
| EU155092.1 | 12 | 2148 | ||
| EU155093.1 | 11 | 2064 | ||
| EU155094.1 | 11 | 2064 | ||
| EU155096.1 | 11 | 1787 | ||
| EU155097.1 | 13 | 2189 | ||
| EU155098.1 | 11 | 2082 | ||
| EU155099.1 | 12 | 2146 |
More Information
Set Membership: |
|

Genomic Map