Assay Details
Target Gene Details
Entrez Gene ID: | 54375 |
Gene Name: | antizyme inhibitor 1 |
Gene Aliases: |
1700085L02Rik, AZI, Oazi, Oazin |
Location: |
Chr.15:38487430-38519287 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Azin1 | NM_001102458.1 | 13 | 3508 | NP_001095928.1 |
| NM_001301688.1 | 12 | 3100 | NP_001288617.1 | |
| NM_018745.5 | 12 | 3348 | NP_061215.1 | |
| NR_125913.1 | 12 | 3497 | ||
| AK049680.1 | 12 | 3349 | BAC33870.1 | |
| AK088671.1 | 13 | 3505 | BAC40494.1 | |
| AK133914.1 | 12 | 3344 |
More Information
Set Membership: |
|

Genomic Map