Assay Details
Target Gene Details
Entrez Gene ID: | 64706 |
Gene Name: | signal peptide, CUB domain, EGF-like 1 |
Gene Aliases: |
7330410C13Rik, A630023E24Rik |
Location: |
Chr.15:83602583-83725049 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scube1 | NM_001271472.1 | 23 | 6303 | NP_001258401.1 |
| NM_001271473.1 | 20 | 5970 | NP_001258402.1 | |
| NM_022723.3 | 22 | 6213 | NP_073560.2 | |
| AK078630.1 | 1 | 832 |
More Information
Set Membership: |
|

Genomic Map