Assay Details
Target Gene Details
Entrez Gene ID: | 53601 |
Gene Name: | protocadherin 12 |
Gene Aliases: |
Pcdh14, VE-cad-2 |
Location: |
Chr.18:38267092-38284401 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdh12 | NM_017378.2 | 4 | 4886 | NP_059074.2 |
| AK028374.1 | 4 | 2002 | ||
| AK090216.1 | 3 | 1706 | ||
| AK165752.1 | 3 | 1706 | ||
| AK165819.1 | 3 | 1706 |
More Information
Set Membership: |
|

Genomic Map