Assay Details
Target Gene Details
Entrez Gene ID: | 66849 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 2 |
Gene Aliases: |
0610025N14Rik, 2310007G06Rik, 4930440J04Rik, 5430408E15Rik, D16Ertd248e, IPP-2 |
Location: |
Chr.16:31251541-31275277 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r2 | NM_025800.3 | 6 | 2477 | NP_080076.1 |
| AK028603.1 | 6 | 2429 | BAC26028.1 | |
| AK032462.1 | 6 | 2428 | BAC27882.1 | |
| AK053657.1 | 6 | 2378 | BAC35465.1 | |
| AK085864.1 | 1 | 415 |
More Information
Set Membership: |
|

Genomic Map