Assay Details
Target Gene Details
Entrez Gene ID: | 24001 |
Gene Name: | T cell lymphoma invasion and metastasis 2 |
Gene Aliases: |
3000002F19Rik, STEF, mKIAA2016 |
Location: |
Chr.17:3326300-3519397 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tiam2 | NM_001122998.1 | 4 | 1727 | NP_001116470.1 |
| NM_011878.2 | 4 | 1711 | NP_036008.2 | |
| AB022915.1 | 4 | 1712 | BAA81823.1 | |
| AK129474.1 | 4 | 1630 | BAC98284.1 | |
| AK158137.1 | 4 | 1690 | BAE34377.1 | |
| BC067048.1 | 4 | 1727 | AAH67048.1 | |
| BC079600.1 | 4 | 1616 | AAH79600.1 |

Genomic Map