Assay Details
Target Gene Details
Entrez Gene ID: | 15547 |
Gene Name: | TRM2 tRNA methyltransferase 2A |
Gene Aliases: |
Htf9c |
Location: |
Chr.16:18248830-18254772 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt2a | NM_001080999.2 | 1 | 116 | NP_001074468.1 |
| NM_001081000.2 | 1 | 116 | NP_001074469.1 | |
| NM_001195205.1 | 1 | 116 | NP_001182134.1 | |
| AK017819.1 | 1 | 127 | ||
| AK034102.1 | ||||
| AK080170.1 | BAC37838.1 | |||
| AK133844.1 | BAE21880.1 | |||
| AK134368.1 | 1 | 117 | BAE22119.1 | |
| AK146391.1 | 1 | 56 | BAE27134.1 | |
| AK150482.1 | BAE29598.1 | |||
| AK151311.1 | BAE30293.1 | |||
| AK151422.1 | BAE30387.1 | |||
| AK151725.1 | BAE30643.1 | |||
| AK151778.1 | BAE30683.1 | |||
| AK152055.1 | BAE30911.1 | |||
| AK152201.1 | BAE31030.1 | |||
| AK152363.1 | BAE31153.1 | |||
| AK152548.1 | BAE31303.1 | |||
| AK152903.1 | BAE31584.1 | |||
| AK153043.1 | BAE31672.1 | |||
| AK153190.1 | BAE31790.1 | |||
| AK169240.1 | ||||
| BC027431.1 | ||||
| BC038486.1 | ||||
| BC046976.1 | AAH46976.1 | |||
| BY090144.1 | ||||
| CJ049472.1 | 1 | 117 | ||
| CX235646.1 | ||||
| X56044.1 | 1 | 320 | CAA39515.1 |
More Information
Set Membership: |
|

Genomic Map