Assay Details
Target Gene Details
Entrez Gene ID: | 74762 |
Gene Name: | MAM domain containing glycosylphosphatidylinositol anchor 1 |
Gene Aliases: |
1200011I03Rik, GPIM, Mamdc3 |
Location: |
Chr.17:29827956-29888429 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdga1 | NM_001081160.1 | 16 | 6086 | NP_001074629.1 |

Genomic Map