Assay Details
Target Gene Details
Entrez Gene ID: | 20405 |
Gene Name: | SH3-domain GRB2-like 1 |
Gene Aliases: |
C77078, EEN, SH3P8, Sh3d2b |
Location: |
Chr.17:56016750-56036638 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3gl1 | NM_001252471.1 | 9 | 1782 | NP_001239400.1 |
| NM_013664.3 | 10 | 1907 | NP_038692.1 | |
| AI846378.1 | 1 | 183 | ||
| AK154836.1 | 10 | 1906 | BAE32864.1 | |
| AK162715.1 | 10 | 1835 | BAE37032.1 | |
| AK167505.1 | 10 | 1849 | BAE39581.1 | |
| CD774556.1 | 1 | 184 | ||
| U58885.1 | 10 | 1846 | AAC71775.1 |

Genomic Map