Assay Details
Target Gene Details
Entrez Gene ID: | 107003 |
Gene Name: | RIKEN cDNA A330093E20 gene |
Gene Aliases: |
AU043608 |
Location: |
Chr.18:45683487-46045260 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A330093E20Rik | NR_040342.1 | |||
| AK039712.1 |
Target Gene Details
Entrez Gene ID: | 140492 |
Gene Name: | potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2 |
Gene Aliases: |
KCa2.2, SK2, SKCA2, fri |
Location: |
Chr.18:45268764-45685887 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnn2 | NM_080465.2 | 8 | 1947 | NP_536713.1 |
| AF357240.1 | 8 | 1947 | AAK48901.1 | |
| AK033158.1 | 11 | 3451 | BAC28176.1 | |
| AK050390.1 | 8 | 2203 | BAC34231.1 | |
| AY123778.1 | 8 | 2022 | AAM88568.1 | |
| BC125475.1 | ||||
| BC137656.1 | ||||
| EU478850.1 | 7 | 1527 |
More Information
Set Membership: |
|

Genomic Map