Assay Details
Target Gene Details
Entrez Gene ID: | 114585 |
Gene Name: | DNA segment, Chr 17, human D6S53E |
Gene Aliases: |
G4, NG34 |
Location: |
Chr.17:35126402-35128855 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D17H6S53E | NM_033477.2 | 3 | 674 | NP_258438.1 |
| BC138707.1 | 1 | 370 | ||
| BC138708.1 | 1 | 370 |

Genomic Map