Assay Details
Target Gene Details
Entrez Gene ID: | 114615 |
Gene Name: | elaC homolog 1 (E. coli) |
Gene Aliases: |
2610018O07Rik, 8430417G19Rik |
Location: |
Chr.18:73735038-73754758 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elac1 | NM_053255.3 | 3 | 481 | NP_444485.2 |
| AF308697.1 | 3 | 480 | AAG24919.1 | |
| CV556353.1 | 1 | 216 |
More Information
Set Membership: |
|

Genomic Map