Assay Details
Target Gene Details
Entrez Gene ID: | 100503704 |
Gene Name: | predicted gene, 16861 |
Gene Aliases: |
- |
Location: |
Chr.16:4935164-4939585 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16861 | NR_037666.1 | |||
| AK078615.2 |
Target Gene Details
Entrez Gene ID: | 17237 |
Gene Name: | mahogunin, ring finger 1 |
Gene Aliases: |
2610042J20Rik, mKIAA0544, md, nc |
Location: |
Chr.16:4886100-4938296 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgrn1 | NM_001252437.1 | 17 | 2825 | NP_001239366.1 |
| NM_029657.4 | 17 | 2822 | NP_083933.1 | |
| AK088533.1 | 17 | 2605 | BAC40408.1 | |
| AK129161.1 | 13 | 2188 | BAC97971.1 | |
| AK189949.1 | 1 | 164 | ||
| AK194956.1 | 1 | 164 | ||
| AK218167.1 | 1 | 164 | ||
| BC046830.1 | 17 | 2671 | AAH46830.1 |
More Information
Set Membership: |
|

Genomic Map