Assay Details
Target Gene Details
Entrez Gene ID: | 52575 |
Gene Name: | tRNA methyltransferase 10C |
Gene Aliases: |
1300018J16Rik, D16Ertd454e, Rg9mtd1, Rnmtd1 |
Location: |
Chr.16:56033720-56037774 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt10c | NM_029092.3 | 1 | 186 | NP_083368.1 |
| AK005047.1 | 1 | 190 | BAB23773.1 | |
| AK087991.1 | 1 | 179 | BAC40080.1 | |
| AK135426.1 | 1 | 192 | BAE22529.1 | |
| AK166781.1 | 1 | 199 | BAE39015.1 | |
| BC023147.1 | 1 | 166 | AAH23147.1 | |
| BC106131.1 | 1 | 168 | AAI06132.1 |
More Information
Set Membership: |
|

Genomic Map